Cell scrapers
Supplier: Biotium
22003
22003.EA 402 EUR
22003.
Cell scrapers
Cell Scrapers
PE, disposable, sterile. Mini cell scrapers for harvesting cells or cell lysates from microwell plates.
Balení: Packaged in 20 sterile packs of 10 scrapers per pack.