Disposable funnel for powders, LaboPlast® / SteriPlast®
Supplier: BURKLE
5378-6005 5378-6006 5379-0041 5379-1041 5378-6007 5378-6008
221-0308EA 17.25 EUR
221-0308 221-0309 221-0310 221-0311 221-0312 221-0313
Disposable funnel for powders, LaboPlast® / SteriPlast®
Funnels Powder Funnels
The disposable powder funnel has a short, wide discharge and is suitable for transferring powdered samples and materials, granulates and suspensions as well as other liquids.
  • For easy and reliable transferring
  • Wide opening prevents clogging
  • Suitable for use in laboratories and clean rooms or for sampling
  • Clean room manufactured (clean room Class 7)
  • Individually packaged and optionally sterilised
  • Complies with EU food requirements and FDA regulations
The wide opening of the disposable powder funnel prevents the powder or granulates from cloggingMore Product Information