Funnels with short stem, DURAN®
Supplier: DWK Life Sciences
213512308 213513304 213513801 213514103 213514609 213515108 213515708 213515905 213516104 213516901 213512805
221-1111EA 13 EUR
221-1111 221-1113 221-1114 221-1115 221-1116 221-1120 221-1117 221-1121 221-1118 221-1119 221-1112
Funnels with short stem, DURAN®
Funnels Filtration Funnels Filter Funnels
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls.
DIN ISO 4798 (exept 221-1112/221-1114/221-1120/221-1118 and 221-1119)
Certifikace: DIN ISO 4798 (exept 221-1112/221-1114/221-1120/221-1118 and 221-1119).