Funnels with extra-long stem
Supplier: KARTELL
162 164 165
221-1860EA 17.1 EUR
221-1860 221-1861 221-1862
Funnels with extra-long stem
Funnels Filtration Funnels Filter Funnels
PP, transparent.
  • Inner ribs to accelerate the filtration process
  • Very good chemical resistance, autoclavable
  • Suitable for use with standard filter papers
With inner ribs for rapid filtration. Cone angle 60°.