Soda lime, AnalaR NORMAPUR® zA, indicator_with

Supplier: VWR Chemicals
Danger

22666.293EA 85.4 EUR
22666.293 22666.362
Soda lime, AnalaR NORMAPUR® zA, indicator_with
Soda lime

For absorbtion of acidic gas (CO₂)


This soda lime contains a new, more efficient saturation indicator. It changes from white to purple when saturated by CO₂

Storage Temperature: Ambient
MDL Number: MFCD00134094
CAS Number: 8006-28-8
Merck Index: 13,08640

Order Now

Specification Test Results

Identification Passes test
Particle size (2 - 5 mm) Min. 78%
Absorption capacity (as CO2) Min. 19%
Loss on drying (105°C) 13 - 18 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR