Hoffmann pinch cocks
Supplier: USBECK Laborgeräte
5510 5511 5512 5513 5514 5515 5516 5520 5521 5522
229-0341EA 5.3 EUR
229-0341 229-0342 229-0343 229-0344 229-0345 229-0346 229-0350 229-0347 229-0348 229-0349
Hoffmann pinch cocks
Clamps Tubing and Sanitary Clamps
Nickel plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Hinged
  • Very good thermal resistance
  • The standard model has a knurled screw and the heavy duty model has a butterfly screw
  • Easy cleaning
Informace pro objednávání: The values in the band width column refer to the clearance between the pinch cock legs.