Nuclear Fast Red, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL

M-Clarity™ quality level = MQ100

229113-5G
229113-5GEA 181 EUR
229113-5G
Nuclear Fast Red, Sigma-Aldrich®
Nuclear Fast Red

Nuclear fast red dye has anthraquinonic nature and is used either alone or in combination with other dyes. It is mainly used for staining of nuclei.


  • Nuclear fast red has been used to counterstain nuclei and renal collecting ducts
  • Suitable for determination of calcium
  • Used as a biological stain

Formula: C₁₄H₈NNaO₇S
MW: 357.28 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00078493
CAS Number: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cb283f0b0d99eea6655bd9e4b7a38026eca4e05&vid=7b686dbb2259750ad6287e881b858a515b7098e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Dark Brown to Very Dark Brown and Dark Red to Very Dark Red and Very Dark Red-Brown
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength 515 - 540 nm
Extinction Coefficient
c = 0.01 g/L, H₂O
≥11000
Suitability
Suitable for Determination of Calcium
Suitable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR