Chemical spoon with spatula end
Supplier: BOCHEM
3421 3422 3423 3424 3425 3750 3420
231-1299EA 4.1 EUR
231-1299 231-1451 231-0240 BOCH3424 BOCH3425 231-2223 231-1298
Chemical spoon with spatula end
Spoons
Stainless steel 18/10, anti-magnetic or stainless steel with PTFE coating. Multipurpose spoon for a wide range of uses.
  • With flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean