Double spatulas, micro
Supplier: BOCHEM
3017 3024 3023 3011 3030 3019 3010 3012 3016 3018 3020 3022 3025 3026 3027 3028 3029 3013 3015 30245 30235 30225 30145 30135 3021 3014
231-0033EA 3.1 EUR
231-0033 231-1027 231-2137 231-0030 231-2242 231-1083 231-0495 231-1016 231-0431 231-0432 231-0541 231-1026 231-0601 231-1029 231-0602 231-0603 231-1084 231-1017 231-0559 231-0037 231-0036 231-0035 231-0032 231-0031 231-0034 231-1018
Double spatulas, micro
Spatulas
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • With round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance