Scoops
Supplier: VITLAB
40093 40193 40293 40393 40493 40593 40693 40793
231-0750EA 9.8 EUR
231-0750 231-0751 231-1086 231-0752 231-1087 231-0753 231-0754 231-0755
Scoops
Scoops
HDPE, translucent. With flat ends and reinforced handle.
  • Good chemical resistance
  • Heat resistant up to +105 °C
  • The capacity of the scoop is stamped into the handle for easy reading