Crucible tongs, stainless steel
Supplier: BOCHEM
2099 2100 2101 2102 2103 2104 2106 2105 2109
231-0182EA 8.9 EUR
231-0182 231-1000 231-1001 231-1002 231-1003 231-1004 231-1006 231-1005 231-0028
Crucible tongs, stainless steel
Tongs
Stainless steel ¹⁸/₁₀, anti-magnetic and electropolished. For easy, safe handling of crucibles made from a wide variety of materials.
  • Double bend with serrated tips
  • Corrosion resistant
  • Excellent thermal resistance, autoclavable