Tissue forceps
Supplier: BOCHEM
1152 1150 1151 1153
232-0003EA 7.1 EUR
232-0003 232-0001 232-0002 232-0004
Tissue forceps
Forceps
Stainless steel.
  • With small teeth (1:2) at the end of the blades
  • Designed for handling tissues with minimal trauma during surgery