Dimethyl sulphoxide, dehydrated (max. 0.03% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

23500.297EA 92 EUR
23500.297 23500.322 23500.260 23500.460
Dimethyl sulphoxide, dehydrated (max. 0.03% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
REACH: 01-2119431362-50
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
IR Spectrum Passes test
Substances coloured by H2SO4 Passes test
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0002 meq/g
Colouration Max. 10 APHA
Melting point Min. 18 °C
Evaporation residue Max. 10 ppm
Heavy metals (as Pb) Max. 1 ppm
Water Max. 0.03 %
Al (Aluminium) Max. 0.2 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 5 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR