Vernier caliper
Supplier: BOCHEM
12110
238-2100EA 92.9 EUR
238-2100
Vernier caliper
Calipers
Stainless steel Vernier caliper with black engraved scale.
Snap length: 45 mm
Fine division: 1/10
Certifikace: DIN 862