Iron(II) sulphate heptahydrate 96, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate

24240.295EA 58.3 EUR
24240.295 24240.364
Iron(II) sulphate heptahydrate 96, GPR RECTAPUR®
Iron (II) sulphate heptahydrate
Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091

Order Now

Specification Test Results

Assay Min. 96 %
Not precipitated by NH4OH (as SO4) Max. 0.5 %
Cl (Chloride) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR