Ammonium iron(II) sulphate hexahydrate 99,0, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Warning

Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate

24257.236EA 67.6 EUR
24257.236 24257.293 24257.260 24257.460
Ammonium iron(II) sulphate hexahydrate 99,0, AnalaR NORMAPUR® zA
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Melting Pt: 110 °C
Density: 1.864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (25°C; 5 %) 3.0 - 5.0
Total P (Phosphorus) Max. 30 ppm
Cl (Chloride) Max. 10 ppm
Ca (Calcium) Max. 10 ppm
Cu (Copper) Max. 10 ppm
Fe (III) (Iron) Max. 0.03 %
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 50 ppm
Mn (Manganese) Max. 0,1 %
Na (Sodium) Max. 50 ppm
Pb (Lead) Max. 10 ppm
Zn (Zinc) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR