Bole white (kaolin), GPR RECTAPUR®, washed

Supplier: VWR Chemicals

Synonyms: China Clay, Hydrated aluminum silicate, Clay

24928.290EA 47.5 EUR
24928.290 24928.321
Bole white (kaolin), GPR RECTAPUR®, washed
Bole white (Kaolin)
Formula: Al₂SiO₇·2H₂O
MW: 258.16 g/mol
Melting Pt: 1760 °C
Density: 0.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS Number: 1332-58-7
EINECS: 310-194-1
Merck Index: 13,05300

Order Now

Specification Test Results

Identity Passes Test
Substances soluble in HCl max. 1.0%
Pb (Lead) max. 0.001%
Loss on ignition (600°C) max. 15.0%
Water max. 1.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR