Magnesium sulphate 98,0, AnalaR NORMAPUR® zA
Supplier: VWR Chemicals
Synonyms: MgSO4, Magnesium sulphate anhydrous
25164.265EA 184 EUR
25164.265
Magnesium sulphate 98,0, AnalaR NORMAPUR® zA
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Min. 98.0 %
Heavy metals (as Pb) Max. 10 ppm
Loss on ignition (600°C) Max. 2 %
Total N (Nitrogen) Max. 40 ppm
Cl (Chloride) Max. 10 ppm
As (Arsenic) Max. 1 ppm
Ca (Calcium) Max. 0.04 %
Fe (Iron) Max. 10 ppm
Mn (Manganese) Max. 20 ppm