Potassium permanganate 99,0-100,5 Ph. Eur.

Supplier: VWR Chemicals
Danger

Synonyms: Permanganic acid potassium salt

26904.293EA 91.5 EUR
26904.293 26904.464
Potassium permanganate 99,0-100,5 Ph. Eur.
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification (K) Passes test
Appearance of solution Passes test
Substances insoluble in water ≤ 1.0 %
Cl (Chloride) ≤ 200 ppm
SO₄ (Sulphate) ≤ 500 ppm
Water ≤ 0.5 %
Residual solvents Passes test
As (Arsenic) ≤ 1 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 1 ppm
Ni (Nickel) ≤ 1 ppm
Pb (Lead) ≤ 1 ppm
V (Vanadium) ≤ 1 ppm
Conforms to Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR