Potassium dihydrogen phosphate 99, GPR RECTAPUR®
Supplier: VWR Chemicals
Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
26923.298EA 69 EUR
26923.298 26923.367
Potassium dihydrogen phosphate 99, GPR RECTAPUR®
Potassium dihydrogen phosphate
Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: ~100 °C (1013 hPa)
Melting Pt: 253
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744
Order Now
Specification Test Results
Assay Min. 99 %
Heavy metals (as Pb) Max. 50 ppm
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 0.02 %
Fe (Iron) Max. 100 ppm