Distilling bridges with Liebig condenser with glass olive or GL thread connection
Supplier: Lenz Laborglas GmbH & CO.KG
05050304 05050404 05050301 05050201 05050211 05050311 05050314
271-3121EA 96.8 EUR
271-3121 271-3122 LENZ05050301 LENZ05050201 LENZ05050211 LENZ05050311 LENZ05050314
Distilling bridges with Liebig condenser with glass olive or GL thread connection
Distillation Components Distillation Bridges
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling links with Liebig condenser, outlet with NS cone, water connection, with socket NS 14/23 for distillation thermometers of 55 mm built-in length.