Sodium acetate trihydrate 99.0-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Supplier: VWR Chemicals

Synonyms: Acetic acid sodium salt trihydrate

27652.232EA 30.8 EUR
27652.232 27652.298 27652.367 27652.260 27652.460
Sodium acetate trihydrate 99.0-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Appearance of solution Passes test Ph.Eur.
Identification A Passes test
Identification B Passes test
IR Spectrum Passes test
Solution in water Passes test
pH (20°C; 5 %) 7.5 - 9.0
pH (25°C; 5 %) 7.5 - 9.0
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Loss on drying (130°C) 39.0 - 40.5 %
Substances reducing KMnO4 (as HCOOH) Max. 50 ppm
Cl (Chloride) Max. 10 ppm
PO4 (Phosphate) Max. 2 ppm
SO4 (Sulphate) Max. 20 ppm
Total N (Nitrogen) Max. 10 ppm
Al (Aluminium) Max. 5 ppm
As (Arsenic) Max. 2 ppm
Ca (Calcium) Max. 10 ppm
Ca + Mg (as Ca) Max. 50 ppm
Cu (Copper) Max. 3 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 5 ppm
Pb (Lead) Max. 5 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR