Sodium chloride ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

27800.291EA 35.4 EUR
27800.291 27800.360 27800.460
Sodium chloride ≥99%, GPR RECTAPUR®
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay Min. 99 %
Hexacyanoferrates (as K4[Fe(CN)6]) Max. 15 ppm
Heavy metals (as Pb) Max. 10 ppm
Br (Bromide) Max. 0.02 %
I (Iodide) Max. 100 ppm
Fe (Iron) Max. 10 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR