Sodium thiosulphate pentahydrate ≥98%, purified

Supplier: VWR Chemicals

27905.293EA 46.4 EUR
27905.293 27905.362
Sodium thiosulphate pentahydrate ≥98%, purified
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Assay Min. 98 %
Heavy metals (as Pb) Max. 20 ppm
Fe (Iron) Max. 20 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR