Sodium iodide 99.0-100.5% Ph. Eur.

Supplier: VWR Chemicals
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

27916.291EA 435 EUR
27916.291 27916.360 27916.462
Sodium iodide 99.0-100.5% Ph. Eur.
Sodium iodide
Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300
Melting Pt: 662
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Order Now

Specification Test Results

Assay (calculated on dried substance) 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
Alkalinity Passes test
IO3 (Iodate) Passes test
SO4 (Sulphate) Max. 150 ppm
S2O3 (Thiosulphate) Passes test
Fe (Iron) Max. 20 ppm
Heavy metals (as Pb) Max. 10 ppm
Loss on drying (105°C) Max. 3 %
Residual solvents Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR