Sodium hydroxide 35% aqueous solution, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Caustic soda

28221.296EA 35.2 EUR
28221.296 28221.365 28221.445 28221.558
Sodium hydroxide 35% aqueous solution, TECHNICAL
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Assay Min. 34 %
Heavy metals (as Pb) Max. 50 ppm
Fe (Iron) Max. 50 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR