Tetrabutylammonium hydrogen sulphate ≥98% for synthesis

Supplier: VWR Chemicals
Warning

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate

28529.186EA 109 EUR
28529.186
Tetrabutylammonium hydrogen sulphate ≥98% for synthesis
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
Melting Pt: 169…172 °C
Density: 1.01 g/cm³ (25 °C)
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5

Order Now

Specification Test Results

Assay Min. 98 %
Melting point 168 - 174 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR