Allylthiourea ≥97%, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea

28630.153EA 88.2 EUR
28630.153
Allylthiourea ≥97%, TECHNICAL
Allylthiourea
Formula: H₂CCHCH₂NHC(S)NH₂
MW: 116.19 g/mol
Boiling Pt: 191 °C (1013 hPa)
Melting Pt: 74 °C
Density: 1.219 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004940
CAS Number: 109-57-9
EINECS: 203-683-5
UN: 2811
ADR: 6.1,III
Merck Index: 12,09500

Order Now

Specification Test Results

Assay Min. 97 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR