p-Xylene ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: 1,4-Dimethylbenzene

28984.292EA 55.4 EUR
28984.292
p-Xylene ≥99%, GPR RECTAPUR®
p-Xylene
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 138.3 °C (1013 hPa)
Melting Pt: 14 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 25
Storage Temperature: Ambient
MDL Number: MFCD00008556
CAS Number: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3 III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b04563a5941cefd103ebd3048bcf8486f2bdac&vid=e5db83ad1f0cb6a583610c58cea291dcbe1be1f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Boiling point 137 - 139 °C
Solidification point 12 - 14 °C
Refractive index (20°C) 1.495 - 1.499
Evaporation residue Max. 50 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR