Zinc, coarse powder, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

29076.267EA 0 EUR
29076.267 29076.291
Zinc, coarse powder, GPR RECTAPUR®
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3 II
Merck Index: 12,10255

Order Now

Specification Test Results

Foreign heavy metals (as Pb) Max. 0.02 %
As (Arsenic) Max. 1 ppm
Fe (Iron) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR