Compressed air dust spray
Supplier: HM INDUSTRIESERVICE
100093 100545
291-1201EA 39.3 EUR
291-1201 HMIN100545
Compressed air dust spray
Sprays
For the quick and easy removal of dust and small amounts of dirt in hard to reach places. For use in medical engineering, metrology, microscopy, precision engineering and in the fields of photography and optics.
  • Includes slip-on capillary tubes (15 cm long) for highly precise spraying
  • Powerful, dry jet of air that is free from oil
  • Non flammable, does not contain any CFCs