Zinc chloride 98,0-100,5, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Supplier: VWR Chemicals
Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

29156.231EA 57.3 EUR
29156.231 29156.297 29156.260 29156.460
Zinc chloride 98,0-100,5, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Zinc chloride
Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 13,10185

Limited Time Sale

Select items below are currently on sale

Order Now

Specification Test Results

Assay 98.0 - 100.5 %
Al + Ca + heavy metals + Fe + Mg Passes test
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
pH (20°C; 10 %) 4.6 - 5.5
Insoluble substances Max. 50 ppm
Oxide chloride (as ZnO) Max. 1.20 %
Total N (Nitrogen) Max. 20 ppm
NH4 (Ammonium) Max. 50 ppm
NO3 (Nitrate) Max. 30 ppm
SO4 (Sulphate) Max. 20 ppm
Ca (Calcium) Max. 10 ppm
Cd (Cadmium) Max. 5 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 0.02 %
Mg (Magnesium) Max. 100 ppm
Na (Sodium) Max. 50 ppm
Pb (Lead) Max. 10 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR