Roller for adhesive seal application
Supplier: Simport Scientific
T329-9
293-1411EA 55.6 EUR
293-1411
Roller for adhesive seal application
Film Applicators Sealing Film Applicators
Ensure a perfect seal when using either SecureSeal™ Thermal sealing film or foil on PCR plates. Roller made of medium-hard rubber.
  • Heavy-duty handle with comfortable grip reducing fatigue
  • Durable construction