Phenol liquified 80% ≥80%

Supplier: VWR Chemicals
Danger

294785AEA 279 EUR
294785A
Phenol liquified 80% ≥80%
Phenol liquified
Formula: C₆H₅OH
MW: 94.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 2821
ADR: 6.1 II

Order Now

Specification Test Results

Assay 78.5 - 81.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR