Disposable foot pump, OTAL®
Supplier: BURKLE
5000-8012
300-0359EA 105.05 EUR
300-0359
Disposable foot pump, OTAL®
Pumps Drum/Transfer Pumps
OTAL® pump with disposable delivery tube made of PE. In OTAL® pumps the medium being transferred only comes in contact with the delivery tube. The disposable delivery tube can be replaced quickly and easily after use.
  • Disposable delivery tube made of PE, can be reordered in a pack of 10
  • Disposable delivery tubes can be cut to length
  • With universal plug made of soft PVC, for container volumes Ø 30 to 70 mm
There is a danger with reusable pumps that the liquid being transferred will be contaminated by the pump, especially if the pump is used forMore Product Information