Steady-Luc™ Firefly HTS assay kit (lyophilised)
Supplier: Biotium
30028-L1 30028-L2 30028-L3
30028-L1EA 151 EUR
30028-L1 30028-L2 30028-L3
Steady-Luc™ Firefly HTS assay kit (lyophilised)
Assays Cellular Assays
Firefly luciferase is widely used as a transcriptional reporter gene for research and drug screening. The Steady-Luc™ HTS kit can be used with luminescence plate readers without sample injection systems or for high-throughput screening.