Sodium selenate decahydrate 97,0, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

302113LEA 192 EUR
302113L
Sodium selenate decahydrate 97,0, GPR RECTAPUR®
Sodium selenate decahydrate
Formula: Na₂SeO₄·10H₂O
MW: 369.09 g/mol
Melting Pt: 35
Density: 1.584 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149174
CAS Number: 10102-23-5
EINECS: 236-501-8
UN: 2630
ADR: 6.1 I

Order Now

Specification Test Results

Assay Min. 97.0 %
Metal impurities Max. 0.02 %
Insolubility in water Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR