Ferroin (1,10-phenanthrolineferrous sulphate complex) (7 mg/ml FeSO₄) aqueous solution, AVS TITRINORM Volumetric, COD_determination_NFT 90-101

Supplier: VWR Chemicals

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate

30890.188EA 51.7 EUR
30890.188
Ferroin (1,10-phenanthrolineferrous sulphate complex) (7 mg/ml FeSO₄) aqueous solution, AVS TITRINORM Volumetric, COD_determination_NFT 90-101
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)

1 ml of solution corresponds to 0.007 g FeSO₄

Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
Density: 0.9999
MDL Number: MFCD00036428
CAS Number: 14634-91-4
EINECS: 238-676-6

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR