Potassium hydroxide 0.05 mol/l (0.05 N) ethanol, AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Danger

Synonyms: Potash caustic, Caustic potash

31305.297EA 67.8 EUR
31305.297 31305.297DP VWRC31305.297_T
Potassium hydroxide 0.05 mol/l (0.05 N) ethanol, AVS TITRINORM Volumetric
Potassium hydroxide

This product may develop a yellow-brown colour during storage.

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1170
ADR: 3 II
Merck Index: 12,07806

Limited Time Sale

Select items below are currently on sale

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR