Centrifuge tubes with plug style caps, UVSafe®
Supplier: LABCON
3132-445-008-9 3142-435-008-9 3142-871-008-9 3182-445-008-9 3192-435-008-9 3192-871-008-9
3132-445-008-9EA 390 EUR
3132-445-008-9 3142-435-008-9 3142-871-008-9 3182-445-008-9 3192-435-008-9 3192-871-008-9
Centrifuge tubes with plug style caps, UVSafe®
Tubes Centrifuge Tubes
Amber UVSafe® centrifuge tubes are tested and lab certified to block 100% of UV wavelengths. Use them instead of wrapping tubes with foil.
  • ATP-, RNase/DNase-, and endotoxin-free (non-pyrogenic)
  • Certified low trace metals (1 ppm)
  • White writing surface
  • Polypropylene USP Class VI resin
  • Free of DiHEMDA, oleamide, and BPA
  • Freezable to –80 °C, and autoclavable
  • Validated Sterilisation (ISO 11137) for use in Research and Medical labs
UVSafe® tubes protect samples, but unlike the opaque black tubes available on the market, they are designed to pass visibleMore Product Information