Sodium hydroxide 0.5 mol/l (0.5 N) aqueous solution, AVS TITRINORM Reag. Ph. Eur. Volumetric
Supplier: VWR Chemicals
Warning
Synonyms: Caustic soda
31951.290EA 19.8 EUR
31951.290 31951.370 31951.404
Sodium hydroxide 0.5 mol/l (0.5 N) aqueous solution, AVS TITRINORM Reag. Ph. Eur. Volumetric
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: >105 °C (1013 hPa)
Melting Pt: 323
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627
Order Now
Specification Test Results
NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l