Drug test kit
Supplier: VWR Chemicals
321481PEA 1120 EUR
321481P 321602G 321612Y 321622K 321632M 321642X 321652Q
Drug test kit
Drug Tests
This kit provides a convenient means of screening suspect material for restricted drugs. The scheme and reagents were developed at the laboratory of the government chemist. Each kit provides sufficient reagents for at least 200 tests.
  • Kit comprises:
  • 20 (4 boxes of 5, 0,2 ml) ampoules of Marquis reagent
  • 1×10 ml cocaine reagent
  • 1×10 ml barbiturate reagent
  • 1×10 ml LSD reagent
  • 1×20 ml cannabis reagent 1
  • 1×10 ml cannabis reagent 2
  • 1×5 g cannabis special solid reagent
  • 1×20 g neutraliser
  • 4 boxes of Whatman filter papers
  • 1 microspatula
  • 20 ampoule breakers
  • 25