Cyanogen bromide

Supplier: Thermo Fisher Scientific
Danger

32731.14 32731.22 32731.06
32731.14EA 65.5 EUR
32731.14 32731.22 32731.06
Cyanogen bromide
Cyanogen bromide

CAS No.: 506-68-3 Molecular Formula: BrCN Formula Weight: 105.93 Physical Form: Crystalline Density: 2.015 Odour: Acrid Boiling Point: 61.4° Merck Reference: 14,2693 UN No.: 1889 MDL No.: MFCD00011597

Formula: CBrN
MW: 105.92 g/mol
Boiling Pt: 61.4 °C
Melting Pt: 50…52 °C
Density: 2.015
Storage Temperature: Refrigerator
MDL Number: MFCD00011597
CAS Number: 506-68-3
EINECS: 208-051-2
UN: 1889
ADR: 6.1 I
Merck Index: 12,02763

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR