Sodium chloride 99.0-100.5% (dried basis), granular USP, FCC, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

Management of Change (MOC) category = HR

3301-37EA 239 EUR
3301-37 3301-39 3301-32 3301-35
Sodium chloride 99.0-100.5% (dried basis), granular USP, FCC, J.T.Baker®
Sodium chloride

Must be subjected to further processing during the preparation of injectable dosage forms.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801 °C
Density: 2.16 g/ml (25 °C)
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity or Alkalinity Passes Test
USP - Appearance of Solution Passes Test
USP - Aluminum (Al) ≤ 0.2 ppm
USP - Arsenic (As) ≤ 1 ppm
USP - Barium (Ba) Passes Test
USP - Bromide (Br) ≤ 100 ppm
USP - Ferrocyanide Passes Test
USP - Iodides (I) Passes Test
USP - Iron (Fe) ≤ 2 ppm
USP - Loss on Drying at 105°C ≤ 0.5 %
USP - Magnesium and Alkaline Earth Metals (as Ca) ≤ 100 ppm
USP - Nitrite (NO₂) (Absorbance Value) ≤ 0.01
USP - Phosphate (PO₄) ≤ 25 ppm
USP - Potassium (K) ≤ 500 ppm
USP - Sulfate (SO₄) ≤ 200 ppm
USP - Assay (NaCl) (dried basis) 99.0 - 100.5 %
FCC - Assay (as NaCl) (FCC) (ignited basis) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Arsenic (As) ≤ 1 ppm
FCC - Calcium and Magnesium ≤ 0.35 %
FCC - Heavy Metals (as Pb) ≤ 2 ppm
FCC - Loss on Drying at 105°C ≤ 0.5 %
Solution Test Passes Test
No Class 1,2,3 or other solvents are used or produced in the ≤1 ppm
manufacturing or purification of the product. ≤0.35%
Must be subjected to further processing during the preparation of ≤2 ppm
injectable dosage forms ≤0.5%
Elemental Impurities (USP 232, EP 5.20) - Information on elemental Passes Test
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR