Sampling spoon, long handle, disposable Bio
Supplier: BURKLE
5379-1021 5379-0021 5379-1022 5379-0022
331-0327EA 25.75 EUR
331-0327 331-0328 331-0329 331-0330
Sampling spoon, long handle, disposable Bio
Spoons
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids. The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
  • Product made from renewable raw materials
  • Bio-PE, white
  • Individually packaged and optionally sterilised
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)