VWR®, Sample Dippers
Supplier: VWR Collection
331-3413EA 424 EUR
331-3413 331-0293 331-0292 331-0294
VWR®, Sample Dippers
Samplers Sample Dippers
All sample dippers are made of blue PP and consist of a bottle with snap handle.
  • Containers designed for sample collection and dispatch for testing in the same container, eliminating risk of cross-contamination
  • Sterilised by electron-beam irradiation and individually packed in sealed zip bag to guarantee integrity of sample
Informace pro objednávání: Available with either hinged plug or screw cap.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904