Diammonium hydrogen citrate 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

33221.22 33221.36 33221.A3
33221.22EA 42.2 EUR
33221.22 33221.36 33221.A3
Diammonium hydrogen citrate 99
di-Ammonium hydrogen citrate

Purity: 99+% CAS No.: 3012-65-5 Molecular Formula: C6H14N2O7 Formula Weight: 226.19 Physical Form: Granular Density: 1.48 Appearance: White Odour: Odourless Merck Reference: 14,512 MDL No.: MFCD00013068

Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
Density: 1.48
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR