Dimethyl glyoxime ≥99% ACS

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime

33310.18 33310.30
33310.18EA 71.2 EUR
33310.18 33310.30
Dimethyl glyoxime ≥99% ACS
Dimethyl glyoxime

CAS No.: 95-45-4 Molecular Formula: C4H8N2O2 Formula Weight: 116.12 Physical Form: Powder Appearance: White to off-white Melting Point: ca 240° dec. Beilstein Reference: 506731 Merck Reference: 14,3246 MDL No.: MFCD00002117

Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Boiling Pt: 53 °C (8 mmHg)
Melting Pt: ∼ 240 °C (dec.)
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
Merck Index: 14,3246

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR