Sodium perborate tetrahydrate, reagent

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sodium peroxometaborate tetrahydrate

33383.14 33383.22 33383.36
33383.14EA 30.8 EUR
33383.14 33383.22 33383.36
Sodium perborate tetrahydrate, reagent
Sodium perborate tetrahydrate

CAS No.: 10486-00-7 Molecular Formula: NaBO3.4H2O Formula Weight: 153.86 (81.84anhy) Physical Form: Granular Density: 1.73 Sensitivity: Hygroscopic Appearance: White Odour: Odourless pH: ca 10 Melting Point: 60° dec. Merck Reference: 14,8652 UN No.: 1479 MDL No.: MFCD00149231


Bleaching agent for domestic detergents and industrial laundries

Formula: NaH₂BO₄·4H₂O
MW: 171.87 g/mol
Melting Pt: 60 °C (dec.)
Density: 1.73
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9
UN: 1479
ADR: 5.1 III
Merck Index: 14,8652

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR