Acetone, J.T.Baker®

Supplier: Avantor

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

3413.5000EA 0 EUR
3413.5000 3413.9010
Acetone, J.T.Baker®
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3 II
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR